Learn More
Medchemexpress LLC Fmoc-Gly(allyl)-OH | 146549-21-5 | 99.9% | 337.38 | 100 MG

Supplier: Medchemexpress LLC HYW008560100MG
Fmoc-Gly(allyl)-OH is a Glycine derivative and an amino acid derivative. It is recognized as a beneficial ergogenic dietary substance, commercially used as an ergogenic supplement influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and prevention of exercise-induced muscle damage.
- Glycine derivative
- Ergogenic supplement for exercise performance
- Influences anabolic hormone secretion
- Supplies fuel during exercise
- Enhances mental performance during stress-related tasks
- Helps prevent exercise-induced muscle damage
- Appears as a white to off-white solid
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage for in-solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.