Learn More
Medchemexpress LLC Fmoc-gly-gly-d-phe-otbu | 98.3% | 557.64 g·mol⁻1 | C32H35N3O6 | 25 MG

Supplier: Medchemexpress LLC HY44234A25MG
Fmoc-Gly-Gly-D-Phe-OtBu is an Fmoc-protected dipeptide-based cleavable linker used as a building block in the synthesis of antibody-drug conjugates (ADCs). It is supplied for research use and represents the R-isomer (D-phenylalanine) configuration.
- Cleavable ADC linker for conjugation and peptide assembly.
- Fmoc-protected N-terminus compatible with solid-phase peptide synthesis.
- D-phenylalanine (R-isomer) configuration for stereochemical control.
- Reported purity approximately 98.3% with enantiomeric excess 99.92% (as listed).
- Molecular formula C32H35N3O6; molecular weight 557.64 g·mol⁻1.
- Available in small-scale quantities for research use only.
- Documentation available: data sheet, certificate of analysis, safety data sheet, handling instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.