Learn More
Medchemexpress LLC Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH | 2264011-98-3 | 100.0% | C33H35N5O9 | 1 ML

Supplier: Medchemexpress LLC HY4554510MM/1ML
Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH is a chemical compound primarily used for synthesizing the ADC Linker, LND1067. This product is intended for research use only.
- High purity of 99.95%
- Chemical formula: C33H35N5O9
- Molecular weight: 645.66
- Appears as a white to off-white solid
- Shipped at room temperature in continental US
- Solid storage at 4°C, sealed and away from moisture
- In-solvent storage at -80°C for up to 6 months or -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.