Learn More
Medchemexpress LLC Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3 | 2866301-96-2 | 98.7% | C33H35N5O8 | 100 MG

Supplier: Medchemexpress LLC HY49412100MG
Fmoc-Gly-Gly-Phe-Gly-NH-CH2-O-CO-CH3 (compound DC-13-C) is an intermediate utilized in the synthesis of Exatecan (HY-13631) derivatives. This product is intended for research use only.
- Appearance: Solid
- Color: White to off-white
- Molecular weight: 629.66
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.