Learn More
Medchemexpress LLC Fmoc-Gly-Gly-Phe-OH | 160036-44-2 | 99.5% | 501.53 g/mol | C28H27N3O6 | 5 G

Supplier: Medchemexpress LLC HY1318335G
Fmoc-Gly-Gly-Phe-OH is an Fmoc-protected tripeptide used as a cleavable linker and intermediate in the synthesis of antibody-drug conjugates (ADCs). It is supplied for research use with supporting product data and safety documentation.
- Cleavable linker for ADC synthesis.
- High purity (99.5%).
- Molecular weight 501.53 g/mol.
- Molecular formula C28H27N3O6.
- Available in multiple pack sizes, including 5 g for synthesis scale-up.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.