Learn More
Medchemexpress LLC Fmoc-Gly-Gly-Phe-OtBu | 236426-37-2 | 99.8% | 557.64 g/mol | C32H35N3O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY4423425MG
Encompass_Preferred
Fmoc-Gly-Gly-Phe-OtBu is an Fmoc-protected tripeptide used as a cleavable linker in antibody-drug conjugate (ADC) synthesis. It provides an Fmoc-protected N-terminus and a tert-butyl ester-protected C-terminus to facilitate peptide coupling and selective deprotection during conjugation workflows.
  • Cleavable linker for ADC synthesis.
  • Fmoc-protected N-terminus for standard peptide coupling.
  • tert-Butyl ester C-terminus for controlled deprotection.
  • High purity suitable for research applications.
  • Available in small-scale quantities for conjugation development.
Catalog No. 50-002-03047