Learn More
Medchemexpress LLC HY-44234 5mg Medchemexpress, Fmoc-Gly-Gly-Phe-OtBu CAS:236426-37-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY442345MG
Encompass_Preferred
Medchemexpress, HY-44234 5mg Fmoc-Gly-Gly-Phe-OtBu CAS:236426-37-2 Fmoc-Gly-Gly-Phe-OtBu is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0065