Learn More
Medchemexpress LLC Fmoc-Gly-NH-CH2-acetyloxy | 1599440-06-8 | 99.7% | 368.38 | 5 G
SDP
Supplier:  Medchemexpress LLC HY449805G
Encompass_Preferred
Fmoc-Gly-NH-CH2-acetyloxy is a cleavable linker that can be used to synthesize antibody-drug conjugates (ADCs).
  • ADCs are comprised of an antibody to which is attached an ADC cytotoxin through an ADC linker.
Catalog No. 50-004-01638