Learn More
Medchemexpress LLC Fmoc-gly-nh-ch2-o-cyclopropane-ch2cooh | 2414254-47-8 | 95.0% | 424.45 | C23H24N2O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW250541100MG
Encompass_Preferred
Fmoc-Gly-NH-CH2-O-Cyclopropane-CH2COOH is an Fmoc-protected glycine-based intermediate for the synthesis of cleavable linkers used in antibody-drug conjugate (ADC) research. Supplied as a solid for research applications, it is intended for use in medicinal chemistry workflows to couple payloads and antibodies.
  • Fmoc-protected glycine scaffold enables standard peptide coupling chemistry.
  • Suitable for synthesis of cleavable ADC linkers and payload conjugation.
  • High purity reported by manufacturer (~95%), appropriate for research use.
  • Solid, white to off-white appearance for straightforward handling.
  • Stable when stored under recommended conditions; in solvent long-term storage at -80 °C.
  • Available in small-scale quantities for medicinal chemistry and linker development.
Catalog No. 50-002-81535