Learn More
Medchemexpress LLC Fmoc-gly-nh-ch2-o-cyclopropane-ch2cooh | 2414254-47-8 | 95.0% | 424.45 g/mol | C23H24N2O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW25054150MG
Encompass_Preferred
Fmoc-Gly-NH-CH2-O-Cyclopropane-CH2COOH is an Fmoc-protected glycine derivative used as an intermediate in the synthesis of antibody-drug conjugate (ADC) linkers. It is supplied as a solid, white to off-white material with a molecular formula of C23H24N2O6 and a molecular weight of 424.45 g/mol. Intended for research use only.
Catalog No. 50-002-81696