Learn More
Medchemexpress LLC Fmoc-gly-nh-ch2-o-cyclopropane-ch2cooh | 2414254-47-8 | 96.0% | 424.45 g/mol | C23H24N2O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW25054125MG
Encompass_Preferred
Fmoc-Gly-NH-CH2-O-Cyclopropane-CH2COOH is an Fmoc-protected glycine derivative used as an intermediate in the synthesis of antibody-drug conjugate (ADC) linkers. It has molecular formula C23H24N2O6, molecular weight 424.45 g/mol, and CAS number 2414254-47-8. The material is supplied for research-scale linker assembly and conjugation workflows, with batch-specific quality documentation available.
  • Used as an intermediate for ADC linker synthesis.
  • Fmoc protection enables peptide coupling and orthogonal deprotection.
  • Batch-specific purity and certificate of analysis available.
  • Storage stability information provided for ambient, refrigerated, and solvent conditions.
  • Available in small quantities appropriate for research and development.
Catalog No. 50-002-81241