Learn More
Medchemexpress LLC Fmoc-Gly-OH-15N (Fmoc-glycine-15N) | 125700-33-6 | 99.7% | 298.30 | 25 MG
Supplier: Medchemexpress LLC 25MGHYY1250S
Fmoc-Gly-OH-15N (Fmoc-glycine-15N) is a 15N labeled, Fmoc-protected glycine derivative, utilized in the synthesis of various compounds. It serves as a valuable tracer and internal standard for quantitative analysis in techniques such as NMR, GC-MS, and LC-MS. The incorporation of stable heavy isotopes like nitrogen-15 into drug molecules is primarily for quantitation during drug development, impacting pharmacokinetic and metabolic profiles.
- Is a 15N labeled, Fmoc-protected glycine derivative
- Used for the synthesis of compounds
- Functions as a tracer for quantitative analysis
- Functions as an internal standard for quantitative analysis
- Applicable in NMR, GC-MS, or LC-MS
- Incorporates stable heavy isotopes into drug molecules for quantitation
- Potential to affect pharmacokinetic and metabolic profiles of drugs