Learn More
Apexbio Technology LLC Fmoc-Gly-OH 29022-11-5 100g
SDP
Supplier:  Apexbio Technology LLC A7901100000
Encompass_Preferred
Fmoc-Gly-OH (29022-11-5) is an Fmoc-protected glycine derivative commonly used in solid-phase peptide synthesis (SPPS) Structurally it features the fluorenylmethyloxycarbonyl (Fmoc) group shielding the amino terminus which can be selectively removed under mild alkaline conditions without compromising peptide integrity This selective deprotection allows for precise stepwise assembly of peptide chains Based on these properties Fmoc-Gly-OH is widely applied in biomedical research including pharmaceutical design peptide synthesis structural characterization and studies on protein function and interactions
Catalog No. 50-000-88660