Learn More
Medchemexpress LLC Fmoc-Gly-OH-d2 (Fmoc-glycine-d2) | 284665-11-8 | 99.5% | 299.32 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYY1250S2
Encompass_Preferred
Fmoc-Gly-OH-2,2-d2 (Fmoc-glycine-2,2-d2) is the deuterium labeled Fmoc-Gly-OH (HY-Y1250). Fmoc-Gly-OH (Fmoc glycine) is a Fmoc-protected glycine derivative, which can be used for the synthesis of compounds. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-93466