Learn More
Medchemexpress LLC Fmoc-Gly-Pro-Hyp(OtBu)-OH TFA | 99.5% | 677.66 | 100 MG

Supplier: Medchemexpress LLC HYP5962100MG
Fmoc-Gly-Pro-Hyp(OtBu)-OH (TFA) is a tripeptide that can be used in peptide synthesis.
- Purity: 99.5%
- Molecular weight: 677.66
- Appearance: Solid
- Color: White to off-white
- Chemical formula: C33H38F3N3O9
- Sequence: Fmoc-Gly-Pro-Hyp(OtBu)-OH
- Store in sealed conditions, away from moisture.
- Powder storage: -80°C for 2 years, -20°C for 1 year.
- In solvent storage: -80°C for 6 months, -20°C for 1 month.
- Solubility in DMSO: 100 mg/mL (147.57 mM)
- For research use only, not intended for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.