Learn More
Medchemexpress LLC Fmoc-Gly-Pro-Hyp(OtBu)-OH (TFA) | 99.5% | 677.66 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP5962
Fmoc-Gly-Pro-Hyp(OtBu)-OH (TFA) is a tripeptide that can be used in peptide synthesis. This product appears as a white to off-white solid with high purity.
- Purity of 99.5%
- Molecular weight of 677.66
- Chemical formula is C33H38F3N3O9
- Appears as a white to off-white solid
- Soluble in DMSO at 100 mg/mL (147.57 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.