Learn More
Medchemexpress LLC Fmoc-Gly-Ser(psi(Me,Me)pro)-OH | 1095952-22-9 | 99.8% | 424.45 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYP2405
Encompass_Preferred
Fmoc-Gly-Ser(psi(Me,Me)pro)-OH is a dipeptide, and it is intended for research use only. This product is offered in various quantities, including 250 mg and 1 g, with bulk inquiry options also available. The product is a solid, white to off-white in color, and should be stored at room temperature in continental US, and sealed away from moisture. For long-term storage as a powder, it can be kept at -80°C for 2 years or -20°C for 1 year. If in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).
  • Dipeptide for research use
  • White to off-white solid appearance
  • Targeted as an amino acid derivative
  • Specific storage conditions apply for powder and in solvent
  • High solubility in DMSO
  • Compatible with various in vivo dissolution protocols
Catalog No. 50-004-26746