Learn More
Medchemexpress LLC Fmoc-Gly-Thr(psi(Me,Me)pro)-OH | 1262308-49-5 | 99.3% | 438.47 | 100 MG

Supplier: Medchemexpress LLC HYP2402100MG
Fmoc-Gly-Thr(psi(Me,Me)pro)-OH is a dipeptide intended for research use only. It is not for sale to patients.
- Purity of 99.3%
- Molecular weight of 438.47
- Solid, white to off-white appearance
- Storage conditions for powder: 2 years at -80°C, 1 year at -20°C
- Storage conditions in solvent: 6 months at -80°C, 1 month at -20°C (sealed storage, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.