Learn More
Medchemexpress LLC Fmoc-Gly-Thr(psi(Me,Me)pro)-OH | 1262308-49-5 | 99.3% | 438.47 | 250 MG

Supplier: Medchemexpress LLC 250MGHYP2402
Fmoc-Gly-Thr(psi(Me,Me)pro)-OH is a dipeptide and a protected amino acid derivative used as a building block in peptide synthesis and other laboratory applications. This product is for research use only.
- High purity: 99.3%
- Appears as a white to off-white solid
- Recommended storage for powder: -80°C for 2 years, -20°C for 1 year
- Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO (250 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.