Learn More
Medchemexpress LLC Fmoc-Gly-Val-OH | 86895-14-9 | 99.8% | 396.44 | 10 G

Supplier: Medchemexpress LLC HYW01375110G
Fmoc-Gly-Val-OH (N-Fmoc-glycyl-L-valine) is a glycyl valine derivative that can be used for the synthesis of drugs or other compounds. It is for research use only and not sold to patients.
- High purity
- White to light yellow solid appearance
- Powder stable for 3 years at -20°C, 2 years at 4°C
- Solution stable for 6 months at -80°C, 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.