Learn More
Bachem Fmoc-His(1-Mtt)-OH
SDP
Supplier:  Bachem 4018533.0005
Encompass_Preferred
Fmoc-His(1-Mtt)-OH, 133367-34-7, C41H35N3O4, Mr 633.75, 5g. Fmoc-His(t-Mtt)-OH. His(Mtt) is somewhat more acid-lable than the Trt derivative. As to minimize racemization, Fmoc-His(Mtt)-OH is best coupled with DEPBT.
Catalog No. 50-312-759