Learn More
Apexbio Technology LLC Fmoc-His(Trt)-OH 109425-51-6 100g
SDP
Supplier:  Apexbio Technology LLC A7904100000
Encompass_Preferred
Fmoc-His Trt -OH (109425-51-6) is a protected amino acid derivative designed for use in solid-phase peptide synthesis The compound features dual protective groups Fmoc (9-fluorenylmethoxycarbonyl) safeguarding the N-terminus and trityl (Trt) protecting the imidazole side chain of histidine This protection strategy enables selective deprotection at defined synthetic steps reducing racemization and minimizing side product formation Based on these chemical properties Fmoc-His Trt -OH is widely utilized in the synthesis of histidine-containing peptides peptide analogs pharmaceutical intermediates and functional biomolecules for biomedical research applications
Catalog No. 50-000-90140