Learn More
Medchemexpress LLC Fmoc-β-HoLeu-OH | 193887-44-4 | MFCD01863059 | 98.4% | 367.44 | 25 G

Supplier: Medchemexpress LLC 25GHYW011089
Fmoc-β-HoLeu-OH is a leucine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder -20°C 3 years, 4°C 2 years; In solvent -80°C 6 months, -20°C 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.