Learn More
Apexbio Technology LLC Fmoc-β-HomoTrp(Boc)-OH 357271-55-7 100g
SDP
Supplier:  Apexbio Technology LLC A6666100000
Encompass_Preferred
Fmoc-beta-HomoTrp Boc -OH is a -amino acid derivative utilized as a building block for peptide modification Incorporation of this compound into peptide sequences introduces structural changes by altering backbone geometry and the spacing between peptide bonds which adjusts peptide secondary structures and binding conformations thereby influencing interactions with biological targets Based on these molecular properties Fmoc-beta-HomoTrp Boc -OH holds research potential in peptide-based drug discovery solid-phase peptide synthesis structure-activity relationship (SAR) studies bioactive peptide screening and synthetic biology applications for novel biomolecule development
Catalog No. 50-000-95017
Only null left
Add to Cart
Request a Quote