Learn More
Apexbio Technology LLC Fmoc-Hyp-OH 88050-17-3 100g
SDP
Supplier:  Apexbio Technology LLC A7599100000
Encompass_Preferred
Fmoc-Hyp-OH (CAS 88050-17-3) is a derivative of hydroxyproline bearing an N-terminal fluorenylmethyloxycarbonyl (Fmoc) protecting group It is designed for the controlled incorporation of hydroxyproline into peptide sequences during solid-phase peptide synthesis The Fmoc group allows for selective deprotection under mild base-mediated conditions Fmoc-Hyp-OH is frequently employed in the construction of collagen-like peptides facilitating studies on the role of hydroxyproline residues in peptide conformation and collagen triple-helix formation It is also utilized in biomaterials research to explore collagen-based scaffolds extracellular matrix interactions and structural mechanisms relevant to tissue engineering and regenerative medicine
Catalog No. 50-000-93614