Learn More
Apexbio Technology LLC Fmoc-Ile-OH 71989-23-6 100g
SDP
Supplier:  Apexbio Technology LLC A7908100000
Encompass_Preferred
Fmoc-Ile-OH (CAS 71989-23-6) is an Fmoc-protected derivative of isoleucine widely utilized as a building block in solid-phase peptide synthesis (SPPS) for the stepwise elongation of peptide chains Fmoc-Ile-OH enables the incorporation of isoleucine residues through selective removal of the Fmoc group under mild basic conditions Based on these properties Fmoc-Ile-OH holds research potential in the synthesis of biologically active peptides for structural functional and activity-based studies including investigations of peptide-protein interactions receptor-ligand binding and bioactivity assessments within cellular and molecular models as well as protein engineering applications
Catalog No. 50-000-94146