Learn More
STA PHARMACEUTICAL US LLC Fmoc-L-Asp(tBu)-L-Thr[PSI(Me,Me)Pro]-OH | 5 g | CAS 920519-32-0 | MDL MFCD18427721
Supplier: STA PHARMACEUTICAL US LLC STAAA112775GM
Fmoc-L-Asp(tBu)-L-Thr[PSI(Me,Me)Pro]-OH is a Amino Acid reagent (Subcategory: Pseudo AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 920519-32-0
- MDL: MFCD18427721
- InChIKey: NKQJRYSIZOXCTJ-LAMXGVLKSA-N
- Molecular Weight: 552.624
- Molecular Formula: C30H36N2O8
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (4S,5R)-3-((S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoyl)-2,2,5-trimethyloxazolidine-4-carboxylic acid
- SMILES: C[C@@H]1[C@@H](C(O)=O)N(C([C@H](CC(OC(C)(C)C)=O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)=O)C(O1)(C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.