Learn More
Cayman Chemical Fmoc-L-Cys mt-OH 10g
SDP
Supplier:  Cayman Chemical 4217710 G
Encompass_Preferred
An amino acid-containing building block; has been used in the solid-phase synthesis of S-palmitoylated peptide antigens for use in macrophage uptake assays and stapled peptide dual agonists of GLP-1R and CCK2 receptors
Catalog No. 50-294-4435