Learn More
STA PHARMACEUTICAL US LLC Fmoc-Cys(SO3Na)-ONa | 5 g | CAS 163558-30-3 | MDL MFCD28134200
Supplier: STA PHARMACEUTICAL US LLC STAAA106885GM
Fmoc-Cys(SO3Na)-ONa is a Amino Acid reagent (Subcategory: Cys) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 163558-30-3
- MDL: MFCD28134200
- InChIKey: BIPPKCSTYKCFHX-SQKCAUCHSA-L
- Molecular Weight: 421.439
- Molecular Formula: C18H15NO7S2(-2)
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29309016
- Country Of Origin: China
- IUPAC: Unable to generate value
- SMILES: O=S(SC[C@@H](C([O-])=O)N=C([O-])OCC1C2=CC=CC=C2C3=CC=CC=C31)(O)=O.[N/A+].[N/A+]
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.