Learn More
Medchemexpress LLC Fmoc-L-cysteic acid 100g
SDP
Supplier:  Medchemexpress LLC HYW097054100G
Encompass_Preferred
Fmoc-L-cysteic acid is an amino acid derivative with an Fmoc protecting group which can be used to synthesize triarylsulfonium compounds for radiolabeling of peptides[1]
Catalog No. 50-002-04759