Learn More
Medchemexpress LLC Fmoc-L-glutamine | 71989-20-3 | 99.15% | 368.38 | 100 G

Supplier: Medchemexpress LLC HYW011084100G
Fmoc-L-glutamine is a glutamine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements and are recognized as beneficial ergogenic dietary substances. They influence the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage.
- Molecular Formula: C20H20N2O5
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US
- Storage: Store at room temperature for 3 years; in solvent at -80°C for 2 years, or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.