Learn More
STA PHARMACEUTICAL US LLC Fmoc-L-hPhe(3-F)-OH | 5 g | CAS 1260594-44-2 | MDL MFCD07372155
Supplier: STA PHARMACEUTICAL US LLC STAAA101135GM
Fmoc-L-hPhe(3-F)-OH is a Amino Acid reagent (Subcategory: Homo-natural AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1260594-44-2
- MDL: MFCD07372155
- InChIKey: USLBDGLIEZPUPZ-QHCPKHFHSA-N
- Molecular Weight: 419.452
- Molecular Formula: C25H22FNO4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(3-fluorophenyl)butanoic acid
- SMILES: O=C(O)[C@H](CCC1=CC=CC(F)=C1)NC(OCC2C3=CC=CC=C3C4=CC=CC=C24)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.