Learn More
Medchemexpress LLC Fmoc-L-lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH | 1662688-20-1 | MFCD34563738 | 99.7% | 1196.51 g·mol^-1 | C64H101N5O16 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW615153250MG
Encompass_Preferred
Fmoc-L-Lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH is an Fmoc-protected lysine derivative used as a building block in peptide chemical synthesis. Supplied as a solid powder and available in solution formulations, it is intended for incorporation into peptide sequences during solid-phase peptide synthesis. Handle using standard laboratory safety procedures.
  • Fmoc-protected N-terminus suitable for Fmoc solid-phase peptide synthesis.
  • OtBu side-chain protecting groups reduce side reactions during assembly.
  • AEEA spacer units provide extended linker functionality on the side chain.
  • Available in multiple package sizes, including 250 mg, and as DMSO solutions.
  • High purity for research-grade peptide synthesis applications.
Catalog No. 50-002-84437