Learn More
Medchemexpress LLC Fmoc-L-lysine hydrochloride | 139262-23-0 | MFCD00190889 | 99.0% | 404.89 g/mol | C21H25ClN2O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW5484771G
Encompass_Preferred
Fmoc-L-lysine hydrochloride is an Fmoc-protected lysine building block used in Fmoc solid-phase peptide synthesis. The hydrochloride salt improves handling and solubility for coupling and deprotection steps. It is commonly used to introduce lysine residues and to prepare modified peptides, including diacylated GLP-1 derivatives with biological activity.
  • Fmoc-protected alpha-lysine suitable for Fmoc solid-phase peptide synthesis.
  • Hydrochloride salt improves solubility and handling.
  • Compatible with standard Fmoc deprotection and coupling chemistries.
  • High purity supports reliable peptide assembly.
  • Useful for incorporation of lysine residues and side-chain modifications.
  • Supplied as a solid for accurate weighing and storage.
Catalog No. 50-002-84476