Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Nle-Wang Resin, 5g, Purity: 100-200 mesh, 1%DVB,0.3-0.8mmol/g
Supplier:  BLD PHARMATECH CO LIMITED BD011427035G
Encompass_Preferred
Fmoc-L-Nle-Wang Resin, 5g, Purity: 100-200 mesh, 1%DVB,0.3-0.8mmol/g
Catalog No. NC3965338