Learn More
Medchemexpress LLC Fmoc-L-photo-leucine | 1360651-24-6 | MFCD31380715 | 99.7% | 365.38 g·mol⁻¹ | C20H19N3O4 | 100 MG

Supplier: Medchemexpress LLC HYW142030100MG
Fmoc-L-photo-leucine is an Fmoc-protected leucine derivative containing a photoreactive diazirine moiety that enables covalent photo-crosslinking when incorporated into peptides or proteins. It is supplied as a high-purity solid for use in solid-phase peptide synthesis, site-specific incorporation into proteins, and photoaffinity labeling to study protein-ligand and protein-protein interactions.
- Contains diazirine photoreactive group for UV-activated crosslinking.
- Suitable for solid-phase peptide synthesis and chemical incorporation.
- Enables photoaffinity labeling and mapping of interaction sites.
- High purity for analytical and preparative applications.
- Soluble in DMSO; ultrasonic assistance recommended.
- Stable when stored at -20°C under inert atmosphere.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.