Learn More
Indofine Chemical Fmoc-L-Serine, 98%, 5 Gm
SDP
Supplier:  Indofine Chemical 041381 5 GM
Encompass_Preferred
FMOC-L-SERINE 5 GM, Amino Acids & Peptides, Fmoc-L-Ser-OH, C18H17NO5, MW: 327.30, >98% by HPLC, 73724-45-5, MFCD00051928
Catalog No. 50-912-576