Learn More
STA PHARMACEUTICAL US LLC Fmoc-L-Tic(6-OH)-OH | 100 g | CAS 936545-85-6 | InChIKey CIRUVUFIOMTUNA-QHCPKHFHSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA10339100GM
Fmoc-L-Tic(6-OH)-OH is a Amino Acid reagent (Subcategory: Aryl AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 936545-85-6
- MDL: No data
- InChIKey: CIRUVUFIOMTUNA-QHCPKHFHSA-N
- Molecular Weight: 415.445
- Molecular Formula: C25H21NO5
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29334990
- Country Of Origin: China
- IUPAC: (S)-2-(((9H-fluoren-9-yl)methoxy)carbonyl)-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
- SMILES: OC([C@@H]1CC2=CC(O)=CC=C2CN1C(OCC3C4=CC=CC=C4C5=CC=CC=C35)=O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.