Learn More
BLD PHARMATECH CO LIMITED Fmoc-L-Trp-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Supplier:  BLD PHARMATECH CO LIMITED BD3024765G
Encompass_Preferred
Fmoc-L-Trp-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965245