Learn More
Medchemexpress LLC Fmoc-L-Val-OH-13C5,15N | 1217442-94-8 | 98% | 345.34 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYI1111S25MG
Encompass_Preferred
Fmoc-L-Val-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-L-Val-OH, a valine derivative for research use. It is not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-31442