Learn More
Medchemexpress LLC Fmoc-leucine-13C6,15N | 1163133-36-5 | MFCD06798058 | 99.9% | 360.36 g/mol | C21H23NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101064S15MG
Encompass_Preferred
Fmoc-leucine-13C6,15N is a stable isotope-labeled N-Fmoc L-leucine supplied as a high-purity solid for analytical and synthetic workflows. It is used as an internal standard, tracer, and building block in mass spectrometry, peptide synthesis, and metabolic labeling studies, enabling accurate quantitation and clear isotopic distinction in complex samples.
  • Stable isotope-labeled with 13C6 and 15N for unambiguous tracing.
  • High purity (≈99.9%) suitable for analytical applications.
  • Optimized for use as an internal standard in LC-MS/MS quantitation.
  • Solid powder form compatible with solid-phase peptide synthesis.
  • CAS number provided for precise chemical identification.
Catalog No. 50-002-12275