Learn More
Medchemexpress LLC Fmoc-lys(5-fitc)-oh | 1095493-05-2 | 95.1% | 757.81 | 100 MG

Supplier: Medchemexpress LLC HY147104100MG
Fmoc-Lys(5-FITC)-OH is a marker used for polypeptides or proteins. This compound utilizes FITC, a fluorescence probe for labeling amines, and is also sensitive to pH and Cu2+.
- Marker for polypeptides or proteins
- Fluorescence probe for labeling amines
- pH-sensitive fluorescence dye
- Cu2+-sensitive fluorescence dye
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.