Learn More
Apexbio Technology LLC Fmoc-Lys(Dnp)-OH 148083-64-1 1g
SDP
Supplier:  Apexbio Technology LLC A79261000
Encompass_Preferred
Fmoc-Lys(Dnp)-OH (CAS 148083-64-1) is a chemically modified lysine derivative featuring an Fmoc protective group at the -amino position and a 2 4-dinitrophenyl (Dnp) substituent on the side chain The Dnp group confers fluorescence quenching properties enabling its use in fluorescence resonance energy transfer (FRET)-based assays Based on these properties Fmoc-Lys(Dnp)-OH holds research potential in enzyme activity analysis protease specificity characterization and synthetic peptide design in biological and biochemical studies
Catalog No. 50-000-90941