Learn More
MilliporeSigma FMOC-LYS(FMOC)-OH
Supplier:  MilliporeSigma 8520410005
Encompass
Fmoc-Lys(Fmoc)-OH; > /=98% by HPLC; White to light yellowish solid crystalline powder; EMD; Novabiochem; 5g; N-alpha,epsilon-di-Fmoc-L-lysine; F.W. 590.7; C36H34N2O6; CAS: 78081-87-5
Catalog No. 50-230-1755