Learn More
Medchemexpress LLC Fmoc-Lys(Mtt)-OH | 167393-62-6 | 99.6% | 624.77 | 10 G

Supplier: Medchemexpress LLC HYW00887210G
Fmoc-Lys(Mtt)-OH is a lysine derivative that influences the secretion of anabolic hormones, the supply of fuel during exercise, mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder -20°C for 3 years, 4°C for 2 years. In solvent -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.