Learn More
Bachem Fmoc-Lys(Mtt)-OH
SDP
Supplier:  Bachem 4025724.0025
Encompass_Preferred
Fmoc-Lys(Mtt)-OH, 167393-62-6, C41H40N2O4, Mr 624.78, 25g. Mtt can be removed selectively from peptides bound to Wang and Rink resin.
Catalog No. 50-258-663