Learn More
Chem-Impex International, Inc. Na-Fmoc-Ne-4-methyltrityl-L-lysine 4-alkoxybenzyl alcohol resin (0.3 - 0.8 meq/g, 200 - 400 mesh) | MFCD07778068 | 5G
Supplier:  Chem-Impex International, Inc. 113575G
Encompass_Preferred
Na-Fmoc-Ne-4-methyltrityl-L-lysine 4-alkoxybenzyl alcohol resin (0.3 - 0.8 meq/g, 200 - 400 mesh), MFCD07778068, 5G
Catalog No. 50-505-865