Learn More
Medchemexpress LLC Fmoc-Lys-OH | 105047-45-8 | 368.43 | 25 G

Supplier: Medchemexpress LLC 25GHYW008176
Fmoc-Lys-OH is a lysine derivative that appears as a solid with a white to yellow color. It is primarily used as a laboratory chemical and in the manufacture of various substances.
- High purity of 99.9%
- Molecular weight of 368.43
- Store at 4°C, sealed, away from moisture
- Soluble in DMSO, methanol, and water
- Suitable for laboratory applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.