Learn More
Sigma Aldrich Fine Chemicals Biosciences Fmoc LysDabcyl OH Novabioc
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 8520960001
Encompass
A modified lysine derivative for the preparation of chromogenically-labeled peptides by Fmoc chemistry.The Dabcyl group quenches the fluorescence of EDANS Mca TET JOE FAM fluorophores making it an extremely useful tool for the synthesis of fluorescence-quenched peptide substrates.The product number for this product was previously 04-12-1236.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-290-8042