Learn More
Medchemexpress LLC Fmoc-Met-OH-13C5,15N | 1217437-64-3 | 377.41 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008371S110MG
Encompass_Preferred
Fmoc-Met-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-Met-OH, which is a Methionine derivative. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules to serve as tracers for quantitation during drug development. Deuteration, in particular, can affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labeled compound
  • Derivative of methionine
  • Used as a tracer for quantitation in drug development
  • For research use only
Catalog No. 50-004-45584